missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Double-Safe™ PFA Safety Coated, Liquid-In-Glass Thermometers: Total Immersion
GREENER_CHOICE

Protect your workplace and ensure a safe environment

30.50 € - 48.65 €

Specifications

Certifications/Compliance Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
For Use With (Application) Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Grade Precision
Material Glass
Thermometer Fill Enviro-Safe™ Liquid
View More Specs

Products 3
Product Code Brand Temperature Range (English) Temperature Range (Metric) Length (Metric) Probe Subdivision Price Quantity & Availability  
Product Code Brand Temperature Range (English) Temperature Range (Metric) Length (Metric) Probe Subdivision Price Quantity & Availability  
13009152
GREENER_CHOICE
View Documents
H-B Instrument™
B60400-1200
-10°C to +110°C 200 mm 1°C
30.50 €
Each
Estimated Shipment: 26-02-2025
to see stock.
Add to basket
 
13069152
GREENER_CHOICE
View Documents
H-B Instrument™
B60400-1800
-20°C to +150°C 300 mm 1°C
48.65 €
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
 
13049152
View Documents
H-B Instrument™
B60400-1600
-20°C to +110°C 300 mm 1°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Environmentally safe thermometers are PFA safety coated to keep liquid and glass contained in the event of breakage
  • Lead-free, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • Manufactured and tested by our USA quality team

Applications:

Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.

Order Info

Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Certifications

Traceable to NIST™. Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.

Specifications

Specifications

Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Precision
Enviro-Safe™ Liquid
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Glass
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.