Learn More
Thermo Scientific™ H-Flex 1 Swinging Bucket Rotor and Accessories
Spin more with less! High capacity and flexibility—all in a small footprint.
Brand: Thermo Scientific™ 75003304
695.01 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
- Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
- Spins blood tubes, conical tubes and microplates without changing rotor or buckets
- Get results faster without the need to change hardware
- Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Specifications
Thermo Scientific H-Flex 1 Rotor 18x15mL Adapter, set/2 | |
Swinging Bucket Rotor | |
4297 | |
Yes |
Molded Polyamide Nylone | |
18 x 15 mL Conical Each Adapter | |
75003300 H-Flex Rotor Cross and 75003301 Buckets | |
Autoclavable |