Learn More
Thermo Scientific™ HAAKE™ RheoWin™ Measuring and Evaluation Software
Product Code. p-7138610 Shop All Thermo Scientific Products
Click to view available options
Type:
Software
For Use With (Equipment):
HAAKE Viscotester iQ
For Use With (Application):
Navigating System Operations
Unit Size:
Each
Product Code. 15375123
Brand: Thermo Scientific™ 0985062
Only null left
Add to basket
Add to basket
Easily navigate your rheometer or viscometer operation using RheoWin Measuring and Evaluation Software.
Easily navigate your rheometer or viscometer operation using the Thermo Scientific™ HAAKE™ RheoWin™ Measuring and Evaluation Software. This user-friendly software for rheometers and viscometers can be customized to offer convenient rheometer or viscometer operation with default settings for beginners and full access to all relevant measuring parameters for advanced rheologists.
The HAAKE RheoWin Measuring and Evaluation Software Features
Customizable Software
  • RheoWin JobManager for fully automated process control ('jobs') of measuring and analysis routines and report printout or export
  • RheoWin DataManager for interactive evaluation of measured data, as well as sophisticated tools for creating reports and generating templates for graphs, tables and screen views
  • RheoWin UserManager for comprehensive management of user access control and assignment of specific access rights
Functionality
  • Manual control for preliminary testing, selected parameters display and saving manually-acquired data
  • Convenient creation and customization of measuring routines using predefined measuring and evaluation elements via 'drag and drop' techniques
  • Fully automated measuring, data evaluation and documentation within one measuring routine
  • Simultaneous measurements using several instruments and data evaluation
  • Freely configurable data export (e.g., ASCII, MS-Excel, XML, etc.)
  • Ability to save graphs in various formats
  • Numerous algorithms for data analysis (e.g., interpolation, regression and automated quality control)
  • Ability to save raw data and numerical values for quality control purposes
  • Integrated image capture with USB and firewire camera
Customization
  • User-defined configuration of paths and subdirectories for data filing
  • Modular generation of a file name and automated saving in a predefined subdirectory
  • Data transfer to ERP and laboratory systems (e.g., SAP™, LIMS, etc.)
  • Online display of all corrections to improve data quality
  • Snapshot for quick characterization of an unknown sample
Operating System:
Windows 10 (32-bit and 64-bit version)
Windows 8.1 (32-bit and 64-bit version)
Windows 8 (32-bit and 64-bit version)
Windows 7 (32-bit and 64-bit version)
Specifications
Type Software
For Use With (Equipment) HAAKE Viscotester iQ
For Use With (Application) Navigating System Operations