Learn More
Thermo Scientific™ HAAKE™ Viscotester™ 3 Rotational Viscometer
Ingenious design makes this portable viscometer the easiest you’ll ever use for fast, reliable measurements anywhere in your process.
Brand:  Thermo Scientific™ 399-0112
 View more versions of this product
Product Code. 16687305
  • 3175.00 € / Each
Explore more special offers
Description
Description
Perform viscosity measurements with ease. The Thermo Scientific™ HAAKE™ Viscotester™ 3 rotational viscometer ensures quick, reliable tests for QC applications.
Use it as a handheld viscometer or on a lab stand. Color coding ensures measuring geometries match the analysis, the display is readable in any lighting, and the viscometer remembers the reference point measurement for you. All that comes in a ready-to-go carrying case.
HAAKE Viscotester 3 Rotational Viscometer is the replacement for the discontinued Thermo Scientific™ HAAKE™ Viscotester™ 1 plus and 2 plus Rotational Viscometers.
Designed for simplicity – The HAAKE Viscotester 3 rotational viscometer is built for ease of use to get you quickly to the right viscosity measurements:
  • Self-explaining handling
  • Smart alignment given by design and visualized by an electronic level
  • Quick coupling of measuring geometries for fast and easy adaptation
  • Color coded geometries and a geometry range indicator ensure correct rotor selection every time
  • Adaptable display brightness ensures ideal readability in any environment
  • Battery and power supply options allow the viscometer to be run anywhere
  • Memory assist functionality allows you to quickly and easily compare measured viscosities with the reference point
  • Selectable viscosity units (mPa s or dPa s)
  • Compatibility with previous viscometer models
Recommended for: your entire process, from accepting the delivery of goods, to sampling during production and quality control (QC).
  • Printing inks, paints, inks and other coatings
  • Shampoos, creams, lotions, and similar personal care products
  • Oil, greases, pastes and similar substances
  • Sauces, thickeners, and similar food products
Specifications
Specifications
HAAKE Viscotester 3 Rotational Viscometer for higher viscosity measurements
± 5% FSD (rotors R1, R2, R3)
± 7.5% FSD (rotor R5)
± 10% FSD (rotor R4)
62.5 rpm
30 to 400 000 mPas
Base viscometer comes in carrying case with instrument holder (for lab stand), one measuring cup (No. 3), three rotors (No. 1, 2 and 3), four rechargeable batteries and one power supply adapter. Easily upgrade your Thermo Scientific HAAKE Viscotester 2 Plus viscometer to this model; the results are compatible.
Operable by battery or power supply
Up to 150°C