missing translation for 'onlineSavingsMsg'
Learn More

Hafnium dichloride oxide octahydrate, 98+% (metals basis excluding Zr), Zr <1.5%, Thermo Scientific Chemicals

Product Code. 11301888
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

For Synthesis of Hafnium (IV) Oxide

Hafnium dichloride oxide octahydrate is used in the synthesis of hafnium(IV) oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14456-34-9
Molecular Formula Cl2H16HfO9
Molecular Weight (g/mol) 409.51
MDL Number MFCD00150417
InChI Key FXQGUVLIMYOGDZ-UHFFFAOYSA-L
Synonym hafnium iv dichloride oxide octahydrate 98+%-hf zr
PubChem CID 131675691
IUPAC Name hafnium;dichloride;nonahydrate
SMILES O.O.O.O.O.O.O.O.Cl[Hf](Cl)=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range >98% (Metals basis excluding Zr)
Solubility Information Soluble in water.
Formula Weight 409.52 (265.40 Anhydrous)
Odor Pungent
Sensitivity Hygroscopic
Density 2.2
Chemical Name or Material Hafnium dichloride oxide octahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'