Learn More
Hafnium dichloride oxide octahydrate, Puratronic™, 99.998% (metals basis excluding Zr), Zr <1%, Thermo Scientific™
75.32 € - 987.00 €
Chemical Identifiers
CAS | 14456-34-9 |
---|---|
Molecular Formula | Cl2H16HfO9 |
Molecular Weight (g/mol) | 409.51 |
MDL Number | MFCD00150417 |
InChI Key | FXQGUVLIMYOGDZ-UHFFFAOYSA-L |
Synonym | hafnium iv dichloride oxide octahydrate 98+%-hf zr |
PubChem CID | 131675691 |
IUPAC Name | hafnium;dichloride;nonahydrate |
SMILES | O.O.O.O.O.O.O.O.Cl[Hf](Cl)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
14456-34-9 | |
409.51 | |
FXQGUVLIMYOGDZ-UHFFFAOYSA-L | |
131675691 | |
O.O.O.O.O.O.O.O.Cl[Hf](Cl)=O |
Cl2H16HfO9 | |
MFCD00150417 | |
hafnium iv dichloride oxide octahydrate 98+%-hf zr | |
hafnium;dichloride;nonahydrate |
Specifications
14456-34-9 | |
Powder | |
HfCl2O·8H2O | |
UN3260 | |
Soluble in water. | |
O.O.O.O.O.O.O.O.Cl[Hf](Cl)=O | |
409.51 | |
409.52 (265.40 Anhydrous) | |
Puratronic™ | |
Hygroscopic | |
(metals basis excluding Zr), Zr <1% |
White | |
Cl2H16HfO9 | |
MFCD00150417 | |
hafnium iv dichloride oxide octahydrate 98+%-hf zr | |
FXQGUVLIMYOGDZ-UHFFFAOYSA-L | |
hafnium;dichloride;nonahydrate | |
131675691 | |
99.998% | |
Pungent | |
2.2 g/mL | |
Hafnium dichloride oxide octahydrate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 237-345-3
missing translation for 'rtecsNumber' : MG4800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.