Learn More
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals
43.92 € - 576.00 €
Chemical Identifiers
CAS | 13499-05-3 |
---|---|
Molecular Formula | Cl4Hf |
Molecular Weight (g/mol) | 320.29 |
MDL Number | MFCD00011034 |
InChI Key | PDPJQWYGJJBYLF-UHFFFAOYSA-J |
Synonym | hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis |
PubChem CID | 37715 |
IUPAC Name | tetrachlorohafnium |
SMILES | Cl[Hf](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351888
|
Thermo Scientific Alfa Aesar
011834.88 |
2.5 g |
43.92 €
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15495985
|
Thermo Scientific Alfa Aesar
011834.14 |
25 g |
71.05 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341888
|
Thermo Scientific Alfa Aesar
011834.22 |
100 g |
222.11 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331888
|
Thermo Scientific Alfa Aesar
011834.36 |
500 g | N/A | ||||||
Description
Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13499-05-3 | |
320.29 | |
PDPJQWYGJJBYLF-UHFFFAOYSA-J | |
37715 | |
Cl[Hf](Cl)(Cl)Cl |
Cl4Hf | |
MFCD00011034 | |
hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis | |
tetrachlorohafnium |
Specifications
320° (sublimation) | |
White | |
>98% (Metals basis excluding Zr) | |
MFCD00011034 | |
Soluble in water,methanol and acetone. | |
Cl[Hf](Cl)(Cl)Cl | |
320.29 | |
320.3 | |
Moisture sensitive |
13499-05-3 | |
Powder | |
Cl4Hf | |
hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis | |
PDPJQWYGJJBYLF-UHFFFAOYSA-J | |
tetrachlorohafnium | |
37715 | |
Irritating | |
Hafnium(IV) chloride |
Safety and Handling
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 236-826-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.