missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) oxide, 98%, Thermo Scientific™

Product Code. 10543941
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Boiling Point 5400°C
Melting Point 2810°C
CAS 12055-23-1
Quantity 1 g
Assay Percent Range 97.5% min
Molecular Formula HfO2
Linear Formula HfO2
MDL Number MFCD00003565
Merck Index 10, 4470
Solubility Information Solubility in water: insoluble
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
SMILES [O--].[O--].[Hf+4]
IUPAC Name hafnium(4+) dioxidandiide
Molecular Weight (g/mol) 210.49
Formula Weight 210.49
Percent Purity 98%
Infrared Spectrum White powder
Packaging Glass bottle
Specific Gravity 9.68
Density 9.6800g/mL
Chemical Name or Material Hafnium(IV) oxide, 98%
Show More Show Less
Product Identifier
  • Hafnium(IV) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'