missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) oxide, 99.9% (metals basis excluding Zr), Zr <0.5%, Thermo Scientific Chemicals

Product Code. 11323248
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hafnium(IV) oxide is an intermediate. Hafnia is used in optical coatings, and as a high-κ dielectric in DRAM capacitors and in advanced metal-oxide-semiconductor devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12055-23-1
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]

Specifications

Melting Point 2774°C
Physical Form Sintered Lump
Quantity 10 g
Merck Index 14,4588
Solubility Information Insoluble in water.
Formula Weight 210.49
Percent Purity 99.9%
Odor Odorless
Density 9.68 g/mL
Assay (metals basis excluding Zr), Zr <0.5%
Chemical Name or Material Hafnium(IV) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.