Learn More
Hafnium(IV) oxide, 99.95%, (metals basis excluding Zr), Zr typically <0.5%, Thermo Scientific Chemicals
38.09 € - 312.56 €
Chemical Identifiers
CAS | 12055-23-1 |
---|---|
Molecular Formula | HfO2 |
Molecular Weight (g/mol) | 210.49 |
MDL Number | MFCD00003565 |
InChI Key | WIHZLLGSGQNAGK-UHFFFAOYSA-N |
IUPAC Name | hafnium(4+) dioxidandiide |
SMILES | [O--].[O--].[Hf+4] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11303827
|
Thermo Scientific Alfa Aesar
035666.03 |
1 g |
38.09 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393817
|
Thermo Scientific Alfa Aesar
035666.06 |
5 g |
139.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11383817
|
Thermo Scientific Alfa Aesar
035666.14 |
25 g |
312.56 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hafnium(IV) oxide is used in optical coatings and in advanced metal-oxide-semiconductor devices. It is used as a gate insulator in field-effect transistors due to its high dielectric constant. It plays an important role as a possible candidate for resistive-switching memories, as a refractory material in the insulation of devices such as thermocouples and as a high-κ dielectric in DRAM capacitors. It is also used in the preparation of hafnium tetrachloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12055-23-1 | |
210.49 | |
WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
[O--].[O--].[Hf+4] |
HfO2 | |
MFCD00003565 | |
hafnium(4+) dioxidandiide |
Specifications
2774°C | |
Powder | |
MFCD00003565 | |
Insoluble in water. | |
[O--].[O--].[Hf+4] | |
210.49 | |
99.95% | |
9.68 g/mL | |
Hafnium(IV) oxide |
12055-23-1 | |
HfO2 | |
14,4588 | |
WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
hafnium(4+) dioxidandiide | |
210.49 | |
Odorless | |
(metals basis excluding Zr), Zr typically <0.5% |
Safety and Handling
missing translation for 'einecsNumber' : 235-013-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.