missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnium(IV) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 1 g |
Assay Percent Range | 99.99% |
Linear Formula | HfO2 |
Formula Weight | 210.49 |
Percent Purity | 85% (Hf typical) |
Grade | Trace Metal Basis |
Packaging | Glass bottle |
Density | 9.6800g/mL |
Show More |
Safety and Handling
Product Identifier
- Hafnium(IV) oxide
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'