missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) sulfate, 99.9% (metals basis excluding Zr), Zr <1%, Thermo Scientific Chemicals

223.00 € - 967.00 €

Chemical Identifiers

CAS 15823-43-5
Molecular Formula HfO4S
Molecular Weight (g/mol) 274.55
MDL Number MFCD00168081
InChI Key KOELDGKLEFTHGN-UHFFFAOYSA-L
Synonym hafnium sulfate, acmc-20aksa, sulfuric acid, hafnium 4+ salt 2:1, hafnium 4+ disulfate, hafnium iv sulfate tetrahydrate, sulfuric acid,hafnium 4+ salt 2:1, hafnium iv sulfate trace metals basis purity excludes zirconium
PubChem CID 13094115
IUPAC Name hafnium(4+);disulfate
SMILES [Hf].[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11399768
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044244.06
5 g
223.00 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11309778
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044244.14
25 g
967.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15823-43-5
274.55
KOELDGKLEFTHGN-UHFFFAOYSA-L
13094115
[Hf].[O-]S([O-])(=O)=O
HfO4S
MFCD00168081
hafnium sulfate, acmc-20aksa, sulfuric acid, hafnium 4+ salt 2:1, hafnium 4+ disulfate, hafnium iv sulfate tetrahydrate, sulfuric acid,hafnium 4+ salt 2:1, hafnium iv sulfate trace metals basis purity excludes zirconium
hafnium(4+);disulfate

Specifications

15823-43-5
HfO4S
hafnium sulfate, acmc-20aksa, sulfuric acid, hafnium 4+ salt 2:1, hafnium 4+ disulfate, hafnium iv sulfate tetrahydrate, sulfuric acid,hafnium 4+ salt 2:1, hafnium iv sulfate trace metals basis purity excludes zirconium
[Hf].[O-]S([O-])(=O)=O
274.55
370.62 (Anhydrous)
(metals basis excluding Zr), Zr <1%
Powder
MFCD00168081
KOELDGKLEFTHGN-UHFFFAOYSA-L
hafnium(4+);disulfate
13094115
99.9%
Hafnium (IV) sulfate
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only