missing translation for 'onlineSavingsMsg'
Learn More

Hafnium silicide, Thermo Scientific Chemicals

Product Code. 11377407
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

They have been used as ohmic contacts, Schottky barrier contacts, gate electrodes, local interconnects, and diffusion barriers. FeSi2 has become the remarkable source as high temperature thermoelectric material. It is also used as a powder in ceramic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12401-56-8
Molecular Formula HfSi2
Molecular Weight (g/mol) 234.66
MDL Number MFCD00167026
InChI Key TWRSDLOICOIGRH-UHFFFAOYSA-N
Synonym hafnium silicide
PubChem CID 11230071
IUPAC Name hafnium;silicon
SMILES [Si].[Si].[Hf]

Specifications

Melting Point 1680°C
Physical Form Powder
Quantity 10 g
Solubility Information it is insoluble in water.
Formula Weight 234.66
Odor Odorless
Chemical Name or Material Hafnium silicide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.