missing translation for 'onlineSavingsMsg'
Learn More

Hafnium sponge, 99.6% (metals basis excluding Zr), Thermo Scientific™

Product Code. p-4408101
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11326617

Brand: Thermo Scientific Alfa Aesar 000674.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-58-6
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
Synonym hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]

Specifications

Color Silver to Gray
Appearance Silver gray sponge
Quantity 10 g
Odor Odorless
Assay Percent Range 99.6%, Zr nominal 3% (Metals basis excluding Zr)
Form Sponge
Chemical Name or Material Hafnium
compliance-icons
Product Identifier
  • Hafnium powder
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
  • Self-heating substance Category 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H251-Self-heating: may catch fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P235+P410-Keep cool. Protect from sunlight.
  • P422-Store contents under .
  • P420-Store away from other materials.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.