missing translation for 'onlineSavingsMsg'
Learn More

Hafnium Standard For ICP, 1000μg/mL in 2% HCl, Assurance Grade, SPEX CertiPrep™

Product Code. 10617704
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Hf1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-58-6, 7647-01-0, 7732-18-5
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]

Specifications

Packaging Triple-leached LDPE bottle
CAS Min % 0.1, 2.0, 97.9
Quantity 125 mL
CAS Max % 0.1, 2.0, 97.9
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
2-5% HCl 1000μg/mL PLHF1-2Y, -2X 10, 000 PLHF1-3Y
Solvent or Matrix 2% HCl
Chemical Name or Material Hafnium Standard
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'