missing translation for 'onlineSavingsMsg'
Learn More

Hafnium wire, 1.57mm (0.0618in) dia, 99.7% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Product Code. 11346768
Click to view available options
Quantity:
25 cm
100 cm
500 cm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-58-6
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
Synonym hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]

Specifications

Quantity 25 cm
Percent Purity 99.7%
Odor Odorless
Weight ≈25.34g/m
Form Wire
Assay (metals basis excluding Zr), Zr nominal 3%
Chemical Name or Material Hafnium wire, 1.57mm (0.0618 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'