missing translation for 'onlineSavingsMsg'
Learn More

Hafnium wire, 1.57mm (0.0618in) dia, 99.7% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

51.56 € - 656.27 €

Chemical Identifiers

CAS 7440-58-6
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
Synonym hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346768
SDS View Documents
Thermo Scientific Alfa Aesar
013435.BU
25 cm
51.56 €
25cm
Please to purchase this item. Need a web account? Register with us today!
11366768
SDS View Documents
Thermo Scientific Alfa Aesar
013435.BY
100 cm
160.80 €
100cm
Please to purchase this item. Need a web account? Register with us today!
11356768
SDS View Documents
Thermo Scientific Alfa Aesar
013435.CC
500 cm
656.27 €
500cm
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
23986
hafnium
Hf
MFCD00011032
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
CHEBI:33343
[Hf]

Specifications

7440-58-6
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
23986
99.7%
≈25.34g/m
(metals basis excluding Zr), Zr nominal 3%
Hf
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
[Hf]
178.49
CHEBI:33343
Odorless
Wire
Hafnium wire, 1.57mm (0.0618 in.) dia.
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-166-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.