missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kleenex™ Hair and Body Shower Gel

Ideal for dispensers in busy washrooms, hotel, and gym bathrooms. Kleenex™ Hair and Body Shower Gel helps maintain hygiene and cleanliness.

Brand:  Kimberly-Clark™ 6332

77.18 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12601376

  • 171.00 € / Pack of 6
Estimated Shipment: 11-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Supplied in packs of 6 x 1 L refill cassettes
  • White shower gel with a luxury feel and subtle fragrance
  • Dispenses up to 1,000 controlled shots of gel
  • Delivers double the amount of washes as conventional gels
  • Simple to install
  • Compatible with Aquarius™ and Kimberly-Clark Professional™ Wall-mounted Soap Dispensers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hair and Body Shower Gel
Gel
6 x 1 L
1000 mL
6.18 kg
White
93 x 93 x 180 mm
33.8 oz.
13.6 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Kleenex™ Hair and Body Shower Gel > Quantity: 6 x 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.