Learn More
HAL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558435
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LALGLVGEGK MWSPKSGWAD AKYVLEAHGL KPVILKPKEG LALINGTQMI TSLGCEAVER ASAIARQADI VAALTLEVLK GTTKAFDTDI HALRPHRG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.
HAL is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. HAL defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids.Specifications
HAL | |
Polyclonal | |
Unconjugated | |
HAL | |
hal; HIS; histidase; histidine ammonia lyase; histidine ammonia-lyase; histidinemia; Hsd; HSTD; Huth; zgc:136980 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3034 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P42357 | |
HAL | |
Recombinant protein corresponding to Human HAL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only