Learn More
3M™ Premium Particulate Respirator
Product Code. 12872191
Click to view available options
:
Filter type: FFP3D; Protection against solid and liquid particulate matter
Product Code. 12872191
Brand: 3M™ 7100081542
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides exceptional comfort because of its lightweight, off-the-face design. The 3M Premium Particulate Respirator has high performance filter material for effective filtration with easy breathing.
  • Exceptional comfort because of its lightweight, off-the-face design
  • Meets the particular requirements of the clogging test for respirators which may be used for more than one shift
  • Premium disposable respirator offers reliable, effective protection against dust, mist, and metal fumes
  • Robust and durable shell and cup design provides multi-shift capability and a securer feel
  • Faceseal can be hygienically cleaned for improved wearer hygiene if used for longer than one shift (using 3M™ Face Seal Cleaner 105)
  • High performance filter material offers effective filtration with easy breathing
  • Increased filter area helps extend the life of the respirator
  • Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 149: FFP3D, European Community Directive 89/686/EEC, CE
Design Type Robust Shell and Cup
Disposable Single-use
Filter Type FFP3 R D
Product Type Premium Particulate Respirator
With Exhalation Valve Yes
For Use With (Application) Protection against dust, mist and Metal fumes
Latex Free Yes
Model 8835
Resistant To Dusts, mists and Metal fumes