Learn More
CleanSpace™ HALO Power System
Battery-powered, positive pressure air-purifying respirator with IP Rating 66. Suitable for healthcare, laboratory, pharmaceutical and emergency response. Protection in applications with particulate contaminants and biohazards as well as heavy water and decontamination procedures.
Brand:  CleanSpace™ CS3000
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15900102
  • 1665.00 € / Each
Explore more special offers
Description
Description
  • IP Rating 66 – resistant to dust and liquid ingress
  • Suitable for decontamination procedures
  • Light weight (350g),compact and comfortable
  • Long operating time: up to 9 hours and quick battery recharging: < 2hours
  • AirSensit™ System for mask pressure control and breath responsive airflow
  • One-button system and standby and auto-start/stop mode for quick don/doffing
  • Easy to use support harness
  • Altitude compensated filter blocked detection
  • Easy and accurate flow capability check
Specifications
Specifications
PAPR Assembly, No Headtop
CS3000
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Mask sold separately