missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erlab™ Halo Sense Smart Pollution Sensor
Erlab™ Halo Sense Smart Pollution Sensor offers continuous filtration and purification of laboratory air.
Description
- Continuous monitoring of laboratory air quality for increased safety for laboratory personnel
- Energy saving possible
- Smart technology informs in real time about a laboratory pollution
- 181 x 181 x 67 mm thick
- Instillation: Fixing via removable plate: wall, bench, ceiling
- eGuard app: Application for remote control and monitoring
- Energy Consumption: 10 W
- Interface: Simple pulsed / audible signal communication (can be deactivated)
- 80 to 240 V, 50/60 Hz
- Connectivity via Ethernet port (RJ45 cable included)
Specifications
2 m cable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title