missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Protease Inhibitor Cocktail (100X)

Inhibit proteolytic degradation during cell lysis and protein extraction with these ready-to-use concentrated stock solutions of protease inhibitors.

134.00 € - 588.00 €

Specifications

Product Type Protease Inhibitor Cocktail
Content And Storage Upon receipt store at 4°C
Form Liquid
Product Line Halt™
Buffer Lysis Buffer
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11834101
View Documents
Thermo Scientific™
78429
5 mL
392.00 €
5mL
Please to purchase this item. Need a web account? Register with us today!
10720825
View Documents
Thermo Scientific™
78430
24 x 100 μL
258.00 €
Each
In Stock
to see stock.
Add to basket
 
11814111
View Documents
Thermo Scientific™
78438
10 mL
588.00 €
10mL
Please to purchase this item. Need a web account? Register with us today!
10516495
View Documents
Thermo Scientific™
87786
1 mL
134.00 €
1mL
In Stock
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ Halt Protease Inhibitor Cocktails are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction procedures.

Halt Protease Inhibitor Cocktails are 100X solutions containing optimized concentrations of six broad-spectrum protease inhibitors stabilized in high-quality dimethylsulfoxide (DMSO). The cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The identity and concentration of each component is fully disclosed, leaving no uncertainties about variables and compatibility with experiments. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively. The stable, concentrated liquid cocktails are easy to use, and the several different package sizes make Halt Protease Inhibitor Cocktails exceptionally convenient for either small-scale or large-scale uses.

Highlights:

  • Better than tablets – the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need

  • No proprietary ingredients – fully disclosed formulation contains AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A in DMSO

  • Multiple package sizes – choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination

  • Two popular formulations – regular formulation includes a separate vial of EDTA solution for optional metalloprotease inhibition; EDTA-free formulation ensures compatibility with isoelectric focusing

  • Lysis buffer compatible – use with Thermo Scientific Pierce™ Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent

Includes:

Protease inhibitor cocktails with EDTA include separate 0.5M EDTA solution.

Specifications

Specifications

Protease Inhibitor Cocktail
Liquid
Lysis Buffer
Protease Inhibitor
Upon receipt store at 4°C
Halt™
Protease Inhibitor
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Halt Protease Inhibitor Cocktail (100X)

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Benzenesulfonylfluoride, 4-(2-aminoethyl)-, hydrochloride (1:1)

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.