missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktail, EDTA-free (100X)

Prevent protein degradation and stabilize phosphorylation with this all-in-one cocktail of protease and phosphatase inhibitors.

307.00 € - 1088.00 €

Specifications

Description Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-Free
Product Type Protease and Phosphatase Inhibitor Cocktail
Form Liquid
Product Line Halt™
Buffer Lysis Buffer
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10127963
View Documents
Thermo Scientific™
78441
1 mL
307.00 €
1mL
In Stock
to see stock.
Edge
Add to basket
 
10311494
View Documents
Thermo Scientific™
78443
24 x 100 μL
423.00 €
Pack of 24
In Stock
to see stock.
Edge
Add to basket
 
11844111
View Documents
Thermo Scientific™
78445
5 x 1 mL
724.00 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
 
10289853
View Documents
Thermo Scientific™
78447
10 mL
1088.00 €
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The all-in-one Thermo Scientific™ Halt Protease and Phosphatase Inhibitor Cocktail provides the convenience of a single solution with full sample protection.

Additional Information and Online Ordering

This broad-spectrum Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases. The 100X cocktails, available in regular and EDTA-free formulations and several package formats, are refrigerator-stable liquids that are very easy to use for nearly any sample or experimental scale.

Highlights:

  • Complete protection – all-in-one cocktail contains both protease and phosphatase inhibitors
  • Multiple package sizes – choose from four convenient packages sizes to match nearly any scale
  • Convenient – the 100X liquid formulation is much easier and more effective to use than individual inhibitors and other formats
  • Easy storage – aqueous-based format allows for convenient cold-room storage without freezing, by contrast with DMSO-based cocktails
  • Compatible – can be used in mass spectrometry (MS) because it does not contain AEBSF, which can cause peaks to shift
  • Validated – fully compatible with Thermo Scientific Pierce™ Cell Lysis Buffers

Includes:

Formulations with and without supplementary vial of EDTA for metalloprotease inhibition, both of which are offered in four package sizes.

Storage Requirements:

Upon receipt store at 4°C. Do not freeze product.

Specifications

Specifications

Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-Free
Liquid
Lysis Buffer
Protease Inhibitor, Phosphatase Inhibitor
Protease and Phosphatase Inhibitor Cocktail
Halt™
Protease Inhibitor, Phosphatase Inhibitor
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Halt Protease & Phosphatase Inhibitor Cocktail EDTA-Free

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

Supplemental information
  • MIXTURE LIST-Contains : trisodium orthovanadate, sodium fluoride

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.