Learn More
MP Biomedicals™ Ham's F-12 (modified) with L-Glutamine, without Sodium Bicarbonate
Ham's F-12 medium was designed for low density, serum-free growth of Chinese Hamster Ovary (CHO) cells. Ham's F-12 is well suited for the growth of primary rat hepatocytes and prostate epithelial cells. It is also the medium of choice for toxicity assays using CHO cells.
Brand: MP Biomedicals™ 091042120
37.29 EUR valid until 2025-12-31
Use promo code "24331" to get your promotional price.
Description
Originally, Ham's Nutrient Mixtures were developed for the clonal growth of several Chinese hamster ovary (CHO) clone cells, Hela cells and mouse L-cells. Both F-10 and F-12 mediums are formulated for use with or without serum, depending on the type of cells being cultured.
Ham's F-12 is well suited for the growth of primary rat hepatocytes and prostate epithelial cells, and is also the medium of choice for toxicity assays using CHO cells. MP Biomedical™ also offers Ham's F-12 supplemented with 25 mM HEPES for improved buffering in the optimum pH range of 7.2 to 7.4.
F12K is a modification of Ham's F-12 and Coon's F-12 with increased amino acid and pyruvate concentrations. Additionally, the salts have been modified (Konigsbergs). F12K has been developed for culturing differentiated rat and chicken cells and primary human liver cells. MgCl2 has been substituted by MgSO4 containing an equivalent amount of Mg ion.
Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Specifications
Cell Culture Medium | |
Glutamine | |
No Sodium Bicarbonate | |
Powder to Make 10 X 1 L Solution |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.