Learn More
Hamilton™ 1700/1000 Series Gastight™ Instrument Syringes: AD Termination
Syringes are designed to fit into, or with, various different instruments such as the MICROLAB™ 500 and MICROLAB 1000
Brand:  Hamilton™ 201000
 View more versions of this product
Product Code. 11557864
  • 120.00 € / Each
Explore more special offers
Description
Description
Have a PTFE plunger tip that creates a tight connection between the syringe barrel and plunger allowing for liquid or gas sampling.
  • AccuDil™ (AD) - This termination is designed for MICROLAB 1000 Diluters/Dispensers
Specifications
Specifications
Borosilicate Glass (Barrel), PTFE (Tip)
1000 psi
50°C
1705
Ea.
AccuDil
Gastight Syringe
122°F
68°F
No Needle Included
50 μL
Microlab 1000 Diluters/Dispensers