Learn More
Hamilton™ 700 Microliter™ Syringes: RN Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
Brand:  Hamilton™ 80330
 View more versions of this product
Product Code. 10363851
  • 110.00 € / Each
Explore more special offers
Description
Description
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hamilton™ 700 Microliter™ Syringes: RN Termination > 10μL; RN; 26s gauge; 2 in. L (51mm); Point style: 2