Learn More
Hamilton™ Chemically Inert Manual Valves
Product Code. p-315949
Click to view available options
Type:
1-1 Manual Valve
3-2 Manual Valve
3-3 Manual Valve
Unit Size:
Each
Product Code. 10337272
Brand: Hamilton™ 86727
Valves have a fluid path containing PTFE and CTFE
The ports on the valve are 1/4-28 threads and come in different flow paths.
Specifications
For Use With (Application) For low pressure applications, from a basic on/off flow to a four-way distribution system
Material CTFE, PTFE
Type 3-3 Manual Valve