Learn More
Hamilton™ Constant Rate-700 HPLC Syringes & Replacement Parts
Syringes are designed to delive reasy, reproducible injection of liquid samples into an injector on HPLC instrumentation. The CR-700 standardizes three major variables: sample volume, rate of injection, and depth of needle penetration. Even when operated by different technicians, the CR-700 provides reproducibility of +1% of the total syringe volume in routine procedures.
Brand:  Hamilton™ 84302
 View more versions of this product
Product Code. 10055610
  • 420.00 € / Each
Explore more special offers
Description
Description
Volumes are selected with micrometer precisions, the plunger is spring-driven and injects the samples at a constant rate. Three models of the CR-700 syringes are available with volumes of 20, 50 or 200μL.
Needle Point Styles
  • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauge: 22
Specifications
Specifications
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
0.016 in.
0.028 in.
2 in.
500 psi
80°C
CR700-200
Ea.
51 mm
HPLC Injection Valves
22 G
0.41 mm
0.72 mm
Gastight Syringe
176°F
50°F
10°C
200 μL
Blunt