Learn More
Hamilton™ Gastight™ Waters WISP Autosampler Syringes
Designed for use with Waters WISP HPLC autosamplers
Brand:  Hamilton™ 80020
 View more versions of this product
Product Code. 11520222
  • 241.00 € / Each
Explore more special offers
Description
Description
Tight fit plunger tip wipes the syringe barrel interior free of sample. This feature is useful with heterogeneous samples as it reduces chances that a deposit will occur and cause plunger to freeze.
  • For dispensing both liquids and gases
  • Precision-machined PTFE plunger tip creates leak-free seal; with the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample
  • Have Cheminert (CX) termination and are designed to fit directly into the instrumentation removing the need for a needle
Specifications
Specifications
Autoclavable
0.305 in.
7.75 mm
1000 psi
115°C
1702C
Ea.
Waters WISP HPLC Autosamplers
0.029 in.
0.72 mm
Gastight Syringe
239°F
50°F
10°C
25 μL