Learn More
Hamilton™ HxSil C18 Guard Cartridge Kit
Stainless steel and PEEK guard columns help protect your investment
Brand:  Hamilton™ 79459
Product Code. 11503254
  • 334.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Guard columns remove particulate contaminants and highly adsorptive compounds from samples
  • Modular and cartridge replacement is easy and tool free
  • Holders are available in stainless steel and PEEK materials, both designs optimized to reduce dead volume and prevent extra column band broadening
  • Holders for the cartridges are reusable indefinitely
Kit contains 1 holder and 2 cartridges
Recommended for use with virtually all solvents except, concentrated nitric acid, sulfuric acid, tetrahydrofuran or dimethylsulfoxide.
Specifications
Specifications
20 mm
C18
Stainless Steel