Learn More
Hamilton™ Metal Hub Blunt Point Needles (Luer Lock)
Can be used with TLL syringes and LT or TLL connectors.
Brand:  Hamilton™ 91023
 View more versions of this product
Product Code. 11537794
  • 94.40 € / Pack of 6
Explore more special offers
Description
Description
Can be used with TLL syringes and LT or TLL connectors.
  • Made of 304 Stainless Steel
  • Standard needle length is 51mm (2 in.)
  • Available in convenient six packs
  • Custom needle lengths and point styles are available
  • Needles are available from 10 to 33 ga
Specifications
Specifications
Autoclavable
23 G
0.025 in.
0.64 mm
Needle
Blunt
Nickel-plated Brass (Hub), Stainless Steel
0.013 in.
0.34 mm
2 in.
51 mm
PTFE Luer-lock Syringes and Luer-tip or PTFE Luer-lock Connectors