Learn More
Hamilton™ Metal Hub Blunt Point Needles (Luer Lock)
Can be used with TLL syringes and LT or TLL connectors.
Brand:  Hamilton™ 91033
 View more versions of this product
Product Code. 11572325
  • 139.00 € / Pack of 6
Explore more special offers
Description
Description
Can be used with TLL syringes and LT or TLL connectors.
  • Made of 304 Stainless Steel
  • Standard needle length is 51mm (2 in.)
  • Available in convenient six packs
  • Custom needle lengths and point styles are available
  • Needles are available from 10 to 33 ga
Specifications
Specifications
Autoclavable
33 G
0.008 in.
0.21 mm
Needle
Blunt
Nickel-plated Brass (Hub), Stainless Steel
0.004 in.
0.11 mm
2 in.
51 mm
PTFE Luer-lock Syringes and Luer-tip or PTFE Luer-lock Connectors