Learn More
Hamilton™ Metal Hub Side Port Needles (Luer Lock)
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors. Needles are 304 stainless steel (sst.). Standard needle length is 2 in. (51mm). Custom needle lengths and point styles are available.
Brand:  Hamilton™ 7729-07
 View more versions of this product
Product Code. 10064664
  • 194.00 € / Pack of 6
Explore more special offers
Description
Description
Metal Hub Compatible Syringe Terminations
  • PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
  • Needle Point Styles
    • Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10
    /li>
    Six needles per pack.
    Specifications
    Specifications
    Autoclavable
    22 G
    0.028 in.
    0.72 mm
    Replacement Needle
    TLL Syringes and LT or TLL Connectors
    Nickel-plated Brass (Hub), Stainless Steel
    0.016 in.
    0.41 mm
    2 in.
    Cone with Side Port