Learn More
Hamilton™ Micro Pipet Disposable Tips
Microsyringe Pipets have special 3/4 in. electrotapered needle for easy attachment of PTFE tips
Brand:  Hamilton™ 84255
Product Code. 11512984
  • 56.40 € / Pack of 100
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Disposable PTFE tips that are to be used with the Microsyringe Pipet (13-684-112A)
  • Sample only wets disposable PTFE tips, eliminating sample cross-contamination
  • Spring-loaded plunger and adjustable stops ensure excellent reproducibility
  • Adjustable stop allows transfer of samples from .2 to 3μL
Specifications
Specifications
PTFE
Single-use
0.2 to 3 μL
Microsyringe Pipet Model 701