Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
Product Code. 15146862
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
Model:
1001 BFP
1002 BFP
1005 BFP
1010 BFP
1701 BFP
1702 BFP
1705 BFP
1710 BFP
1725 BFP
1750 BFP
Bubble free prime
Unit Size:
Each
Product Code. 15146862
Brand: Hamilton™ 5900010
For use with MICROLAB 600 Diluter/Dispenser
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Specifications
Model 1702 BFP
Volume (Metric) 25 μL
For Use With (Equipment) Microlab™ 600 dispensers