missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Gastight™ Removable Autosampler Syringes For Agilent 7670, 7671, and 7672 ALS GC
Syringes are designed to work with Agilent 7670, 7671, and 7672 Automatic Liquid Sampler (ALS) gas chromatography (GC) autosamplers
Marque: Hamilton™ 80011
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Syringes in this category are either 700 series Microliter or 1700 series Gastight syringes. Microliter syringes have a stainless steel plunger that is hand fitted to the glass barrel and are designed for liquid sampling. Gastight syringes have a PTFE plunger tip that creates a tight connection between the syringe barrel and the plunger, allowing for liquid or gas sampling.
- Removable Needle (RN) - The needles are removable and are a Hamilton-specific design
- The design allows the needles to seat precisely at the zero graduation mark of the syringe
- Users can select the needle gauge, length, and point style to optimize the syringe for custom applications
- Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging
- Autoclavable when disassembled; repeated autoclaving will shorten syringe life
- Needle Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration
- Available gauge: 26s
Spécification
Borosilicate Glass (Barrel), Stainless Steel (Plunger) | |
0.018 in. | |
0.013 cm | |
0.26 in. | |
0.66 cm | |
2 in. | |
51 mm | |
68.9 bar | |
115°C | |
1701 N | |
10°C | |
10 μL | |
Agilent 7670, 7671, and 7672 Automatic Liquid Sampler, Gas Chromatography Autosamplers |
26s G | |
0.005 in. | |
0.018 in. | |
0.046 cm | |
0.047 cm | |
1.71 in. | |
Gastight Autosampler Syringe | |
239°F | |
50°F | |
2 | |
Ea. | |
Sharp, Bevel, Curved, Non-coring | |
Removable |