Learn More
Kinematica™ Hammer Grinding Attachment for Polymix™ MFC 90 D Mill
Consists of a rotor with three hardened strike hammers and a grooved stator. During the grinding process, the sample is spun around the grinding chamber and struck until the desired particle size has been achieved. The ground sample passes through the sieve into the collection container.
Brand:  Kinematica™ 35031001
Product Code. 11920359
  • 692.00 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Order Info
Includes
Specifications
Specifications
Hammer Grinding Attachment
PX-MFC 90 D
3-armed rotor with cutting knives, stator with 3 cutting bars, and tool