missing translation for 'onlineSavingsMsg'
Learn More

Hammond™ Drierite™ Desiccant Bags, Cole-Parmer™

Product Code. 11832692
Click to view available options
Quantity:
3 g
6 g
1 oz.
8 oz.
This item is not returnable. View return policy
This item is not returnable. View return policy

Protect materials from corrosion with these desiccant bags. Hammond™ Drierite™ Desiccant Bags reduce the humidity to a 100°F dew point in 10 hours and are dust-tight. Materials packaged with these bags are protected from corrosion for many months.

Nonwoven crepe bags filled with Drierite (anyhydrous calcium sulfate) completely dry the air within a package or container during shipment or storage.
  • Sturdy and held together by heat seals
  • Perform well in elevated temperatures
  • Will not release absorbed moisture at less than 350°F

Specifications

Material Calcium Sulfate
Quantity 3 g
Suitability These Bags are Dust-Tight and Will Protect For Many Months
compliance-icons
Product Identifier
  • X750 BAG DRIERITE 3G
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contains : calcium sulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.