missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Scott™ Luxury Foam Frequent Use Hand Cleanser

Product Code. p-8005909
Click to view available options
Volume (Metric):
1000 mL
1200 mL
Unit Size:
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 12621356

Brand: Kimberly-Clark™ 6345

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Dispenses the ideal amount of soap for each wash preventing overuse. Scott™ Luxury Foam Frequent Use Hand Cleanser delivers up to twice as many washes per liter as conventional liquid soaps.

  • Color-free and fragrance-free foam
  • Luxuriant feel formulated for frequent use
  • Strong, hygienically sealed cassettes that are quick and easy to install
  • Give up to 2,500 shots of soap
  • Cassettes shrink to save space when empty and are recyclable (after the pump is removed)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Cleanser
Color Clear
Quantity 4 Electronic Cassettes
Size Up to 2,500 shots
Volume (English) 40.6 oz.
Volume (Metric) 1200 mL
Weight (Metric) 4.8 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.