missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kleenex™ Ultra™ Hand Cleanser Cassette

Kleenex™ Ultra Hand Cleanser Cassette is a solution to help maintain the highest standards of hygiene and cleanliness.

Brand:  Kimberly-Clark™ 6330

80.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10640863

  • 197.00 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Amber
  • Richly lathering liquid hand soap with a clean, floral scent, containing aloe vera
  • Each cassette delivers up to 1,000 shots of soap making it ideal for everyday use, providing a high-quality feel whilst managing costs and reducing waste
  • When the cassette is empty, it can be compressed to conserve space and is fully recyclable once the pump is removed
  • For busy washrooms
  • Professional solution to help maintain the highest standards of hygiene and cleanliness
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hand Cleanser
Liquid
6 x 1 L
1 L
Amber
For use in any washroom environment
6 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Kleenex™ Ultra™ Hand Cleanser Cassette > Capacity: 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.