Learn More
Invitrogen™ Hand-Held Magnetic Plate Washer
Designed for use with the ProcartaPlex assay configured for magnetic separation. The handheld magnetic separator enables you to remove the liquid contents of the 96-well plate by simply decanting or flicking the contents into a sink and blotting off the remainder on a paper towel.
Brand:  Invitrogen™ EPX-55555-000
Product Code. 15303225
  • 1232.00 € / Each
Explore more special offers
Description
Description
  • Reactivity: Canine, Human, Monkey, Mouse, Pig, Rat
  • Please refer to the appropriate ProcartaPlex User Manual for details on running the assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Multiplex Immunoassay
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Hand-Held Magnetic Plate Washer > Magnetic Plate Washer