missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Hand Operated Easy Pump

Sampling Systems™ Hand Operated Easy Pump is ideal for many laboratory applications where small to medium quantities of liquid need to be pumped.

Brand:  Sampling Systems™ 5056A-18

Product Code. 12848161

  • 139.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Rigid polypropylene construction
  • Supplied with a Universal Bung to allow pumping with any container that has a round opening between 30 and 70mm
  • Maximum length of dip tube: 700mm
  • Designed to be easy to install, easy to use, and easy to clean
  • Capacity of 26L/min
  • Tube diameter of 18mm
Specifications

Specifications

700 mm
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.