missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Hand Operated Vacuum Pumps

Produce up to 25 in Hg vacuum or test for pressure leaks with this vacuum pump. Cole-Parmer™ Hand Operated Vacuum Pumps are sealed and self-lubricated for a long, maintenance-free life.

Brand:  Cole-Parmer™ 79301-21

194.75 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 11705199

  • 205.00 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Vacuum gauge is graduated in both centimeters and inches of mercury
  • Built-in trigger enables you to release vacuum without disconnecting the pump
  • Zinc alloy is perfect for rugged applications; PVC is lightweight and easy to handle
Specifications

Specifications

190.5 x 50.8 x 241.3 mm
PVC
35 mL/stroke
Hand Operated Vacuum Pump (without gauge)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.