missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Hand-Shovel
The Hand Shovel is designed for spreading and picking up granules.
22.00 € - 36.00 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
17876416
|
asecos™
14369 |
187.00 mm |
22.00 €
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
17866406
|
asecos™
14370 |
240.00 mm |
36.00 €
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
Description
- Hand shovel for granules.
- PP
- red
Specifications
Polypropylene | |
Shovel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title