missing translation for 'onlineSavingsMsg'
Learn More

Brady™ HandiMark™ Vinyl Repositionable Film

45.68 € - 86.99 €

Specifications

Description HandiMark Repositionable Vinyl Film
Adhesive Type Acrylic, Repositionable
Finish Gloss
Material Vinyl
Product Type Repositionable Film
View More Specs

Products 6
Product Code Brand Color Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Color Length (Metric) Width (Metric) Price Quantity & Availability  
12880590
View Documents
Brady™
042100
White 15 m 13 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12800600
View Documents
Brady™
042102
White 15 m 50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12890590
View Documents
Brady™
042101
White 15 m 25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12820600
View Documents
Brady™
042104
Yellow 15 m 25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12810600
View Documents
Brady™
042103
Yellow 15 m 13 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10001734
View Documents
Brady™
042105
Yellow 15 m 50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

HandiMark Repositionable Vinyl Film
Gloss
Repositionable Film
HandiMark
-40°C to +82°C
15 m
Acrylic, Repositionable
Vinyl
For temporary, removable and repositionable labels, like 5S and lean inventory labels
1 Roll
0.031 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.