missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile
Black tips for accurate sample delivery
Brand: Axygen™ TT-50-CBK-HTR-36
This item is not returnable.
View return policy
Description
- Polypropylene
- Volume ranging from 20 to 1000μL
- Available with or without aerosol-resistant filters
- Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems
Certifications
Certified DNAse-, RNAse- and Pyrogen-safe
Specifications
50 μL | |
Non-sterile | |
Autoclavable | |
For accurate sample delivery | |
Automation, Round | |
Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head |
Non-filtered | |
Black | |
Non-ANSI | |
Rack | |
Hanging Tip Racked, Cardboard pack | |
Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile > Vol.: 50Glc muL; Tip color: Black; Non-sterile; Non-filtered; Cardboard pack
Spot an opportunity for improvement?Share a Content Correction