missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Checker™ pH Tester and Replacement Part

For use with virtually any electrode that shares same connector.

Brand:  Hanna Instruments™ CHECKER1

Product Code. 12333408

  • 321.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

 

  • pH range from 0.00 to 14.00
  • Provides high accuracy with 0.1 pH resolution 
  • 1.5V
  • RH max 95%
  • With LCD and manual two point calibration
  • 3000 hours of battery life

 

Specifications

Specifications

pH
±0.2pH
0 to 14 pH
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers