missing translation for 'onlineSavingsMsg'
Learn More

Hanus solution, Honeywell Fluka™

Product Code. 15634800
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, 0.1 M IBr

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-33-5
Molecular Formula BrI
Molecular Weight (g/mol) 206.81
MDL Number MFCD00011353
InChI Key CBEQRNSPHCCXSH-UHFFFAOYSA-N
Synonym iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
PubChem CID 82238
IUPAC Name iodobromane
SMILES BrI

Specifications

Quantity 6 x 1 L
UN Number UN2789
Flash Point 40°C (104°F)
Formula Weight 206.81
Chemical Name or Material Hanus solution
compliance-icons compliance-icons
Product Identifier
  • Hanus solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Acetic acid, Iodinebromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.