Learn More
3M™ Hard Hat with Plastic Sweatband, G2000CUV-VI
3M™ Hard Hat G2000CUV-VI features a ventilation inlet on the top and a ventilated plastic headband to provide comfort.
Brand:  3M™ 7000039611
Product Code. 15453355
  • 18.00 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Lightweight white hard hat with pinlock suspension
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV radiation and it's time to replace it
  • Extended edge on the back of the hard hat provides extra protection for the sensitive neck area
  • Flat surface for printing a brand name or logo
  • Easy to replace sweatband for comfort and hygiene
  • Ventilated headband for improved comfort
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN397
UV-Stabilized ABS
340 g
White
54 to 62 cm
Ventilated